Anti-EGFR (Ab-1) (528) Mouse Monoclonal Antibody [clone: 528]
Catalog # GR01-100
Supplier: Merck Millipore (Oncogene)
Specifications
- Antibody type:Primary
- Antigen symbol:EGFR
- Clonality:Monoclonal
- Clone:528
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Neutralisation:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: EGFR (Ab-1) (528)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human