Order Entry
Denmark
ContactUsLinkComponent
Anti-Actin (Ab-1) (JLA20) Mouse Monoclonal Antibody
Anti-Actin (Ab-1) (JLA20) Mouse Monoclonal Antibody
Catalog # CP01-1
Anti-Actin (Ab-1) (JLA20) Mouse Monoclonal Antibody
Catalog # CP01-1

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Actin
  • Antigen symbol:
    ACT1
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Multiple
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    1 Each
  • Pk:
    1

Specifications

About this item

Type: Primary
Antigen: Actin (Ab-1) (JLA20)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Pan