Anti-UCP2 Rabbit Polyclonal Antibody
Catalog # 662047-100
Supplier: Merck Millipore (Oncogene)
Specifications
- Antibody type:Primary
- Antigen name:Uncoupling protein 2
- Antigen symbol:UCP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: UCP2 (uncoupling protein 2 (mitochondrial, proton carrier))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: