Order Entry
Denmark
ContactUsLinkComponent
Anti-PAI-1 (MA-33H1F7) Mouse Monoclonal Antibody [clone: MA-33H1F7]
Anti-PAI-1 (MA-33H1F7) Mouse Monoclonal Antibody [clone: MA-33H1F7]
Catalog # 528217-100
Anti-PAI-1 (MA-33H1F7) Mouse Monoclonal Antibody [clone: MA-33H1F7]
Catalog # 528217-100

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    PAI-1
  • Clonality:
    Monoclonal
  • Clone:
    MA-33H1F7
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Neutralisation:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: PAI-1 (MA-33H1F7)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: