Order Entry
Denmark
ContactUsLinkComponent
Anti-Hsp70 (C92F3A-5) Mouse Monoclonal Antibody [clone: C92F3A-5]
Anti-Hsp70 (C92F3A-5) Mouse Monoclonal Antibody [clone: C92F3A-5]
  386032-50
Anti-Hsp70 (C92F3A-5) Mouse Monoclonal Antibody [clone: C92F3A-5]
  386032-50

 

  • Antibody type:
    Primary
  • Antigen symbol:
    HSP70
  • Clonality:
    Monoclonal
  • Clone:
    C92F3A-5
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Multiple
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µg
  • Pk:
    50 µG

 

 

Type: Primary
Antigen: Hsp70 (C92F3A-5)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Pan