Order Entry
Denmark
ContactUsLinkComponent
Anti-G q alpha- and Rabbit Polyclonal Antibody
Anti-G q alpha- and Rabbit Polyclonal Antibody
  371751-50
Anti-G q alpha- and Rabbit Polyclonal Antibody
  371751-50

 

  • Antibody type:
    Primary
  • Antigen name:
    G q alpha- and
  • Antigen symbol:
    GQ
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Western blot:
    Yes
  • Antigen synonyms:
    11A-SUB
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µl
  • Pk:
    50 µl

 

 

Type: Primary
Antigen: G q alpha- and
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: