Ordreindtastning
Denmark
ContactUsLinkComponent
Anti-PTK2 Rabbit Polyclonal Antibody
Anti-PTK2 Rabbit Polyclonal Antibody
Catalog # 341292-10
Anti-PTK2 Rabbit Polyclonal Antibody
Catalog # 341292-10

Specifikationer

  • Antistof type:
    Primary
  • Antigen name:
    protein tyrosine kinase 2
  • Antigen symbol:
    PTK2
  • Clonality:
    Polyclonal
  • Konjugation:
    Unconjugated
  • Vært:
    Rabbit
  • ImmunFluorescens:
    Yes
  • Western blot:
    Yes
  • Opbevaringstemperatur:
    Cool
  • Shipping temperature:
    Ice
  • Størrelse:
    10 Tests
  • Pakn:
    10 Tests

Specifikationer

Om denne vare

Type: Primary
Antigen: PTK2 (protein tyrosine kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: