Anti-CANX Rabbit Polyclonal Antibody
208880-50
- Antibody type:Primary
- Antigen name:calnexin
- Antigen symbol:CANX
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Amino acid number:575 to 593
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Type: Primary
Antigen: CANX (calnexin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: