Order Entry
Denmark
Orders LinkContactUsLinkComponent
Anti-Complement 3 Goat Polyclonal Antibody

Anti-Complement 3 Goat Polyclonal Antibody

1 ml
Catalog # 204869-1
Anti-Complement 3 Goat Polyclonal Antibody
Catalog # 204869-1
Anti-Complement 3 Goat Polyclonal Antibody

1 ml

Price
1.460,00 kr
Log in for availability
1 * 1 mL
Product Details & Documents

Type: Primary
Antigen: Complement 3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:

Specifications
  • Size:
    1 ml
  • Western blot:
    Yes
  • Cat. No.:
    204869-1
  • Clonality:
    Polyclonal
  • Flow cytometry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Antibody type:
    Primary
  • ImmunoFluorescence:
    Yes
  • Storage temperature:
    Cool
  • Host:
    Goat
  • Conjugation:
    Unconjugated
  • Shipping temperature:
    Ice
  • Antigen name:
    Complement 3