Anti-Centromere protein F / mitosin Goat polyclonal antibody unconjugated
Catalog # EVBIEB05622
Supplier: Everest Biotech
Specifications
- Antibody type:Primary
- Antigen name:Centromere Protein F, 350/400kDa
- Antigen symbol:CENPF
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Bovine
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Antigen synonyms:mitosin
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: Centromere protein F / mitosin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Bovine, Human