Anti-SNAP25 Rabbit Polyclonal Antibody
Catalog # BSBTPA1315
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Synaptosomal-Associated Protein, 25kDa
- Antigen symbol:SNAP25
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:FLJ23079|synaptosomal-associated protein 25|SUP|SEC9|synaptosomal-associated 25 kDa protein|bA416N4.2|dJ1068F16.2|SNAP|SNAP-25|RIC-4|RIC4|resistance to inhibitors of cholinesterase 4 homolog|super protein
- Amino acid number:186 to 200
- Sequence:DSNKTRIDEANQRAT
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human SNAP25 (186-200aa DSNKTRIDEANQRAT), identical to the related rat and mouse sequences.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Synaptosomal-associated protein 25(SNAP25) detection. Tested with WB, IHC-P in Human; Mouse; Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: SNAP25
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat