Anti-NOS1 Mouse Monoclonal Antibody [clone: N1]
Catalog # BSBTMA1072
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Nitric Oxide Synthase 1
- Antigen symbol:NOS1
- Clonality:Monoclonal
- Clone:N1
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Pig,Goat,Human,Rat
- Western blot:Yes
- Cross adsorption:No
- Format:Ascites
- Form:Lyophilized
- Antigen synonyms:N-NOS|Peptidyl-cysteine S-nitrosylase NOS1|Constitutive NOS|NC-NOS|Nitric oxide synthase|bNOS|brain|NOS1|NOS type I|nNOS|Neuronal NOS
- Amino acid number:1 to 181
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:Recombinant neuronal NOS fragment (amino acids 1-181) from rat brain.
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: NOS1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat, Goat, Pig