Anti-MAS1 Rabbit Polyclonal Antibody (DyLight® 405)
Catalog # NOVUNLS1531V
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:MAS1 proto-oncogene, G protein-coupled receptor
- Antigen symbol:MAS1
- Clonality:Polyclonal
- Conjugation:DyLight® 405
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Mouse
- Gene ID:4142
- Antigen synonyms:MGRA|MAS
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 μL
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: MAS1 (MAS1 oncogene)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse