Anti-GPR39 Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
Certificates
About this item
Rabbit Polyclonal GPR39 Antibody. Tested Applications: Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:100
Type: Primary
Antigen: GPR39
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Specifications
- Catalog No:
- NOVUNLS139SS
- NOVUNLS139
- Antigen symbol:
- GPR39
- GPR39
- Antigen name:
- G Protein-Coupled Receptor 39
- G Protein-Coupled Receptor 39
- Antigen synonyms:
- G-protein coupled receptor 39|GPR39
- G-protein coupled receptor 39|GPR39
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 2863
- 2863
- Western blot:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together