Order Entry
Denmark
ContactUsLinkComponent
 
Anti-GLP1R Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
  NOVUNLS1205H
Anti-GLP1R Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
  NOVUNLS1205H

 

  • Antibody type:
    Primary
  • Antigen name:
    Glucagon-like peptide 1 receptor
  • Antigen symbol:
    GLP1R
  • Clonality:
    Polyclonal
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Gene ID:
    2740
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 μL
  • Pk:
    50 µl

 

 

Type: Primary
Antigen: GLP1R (glucagon-like peptide 1 receptor)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse