Anti-TLR10 Mouse Monoclonal Antibody [clone: 3C10C5]
Catalog # NOVUNBP2-27214
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Toll-like Receptor 10
- Antigen symbol:TLR10
- Clonality:Monoclonal
- Clone:3C10C5
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Gene ID:81793
- Antigen synonyms:CD290
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: TLR10 (toll-like receptor 10)
Clonality: monoclonal
Clone: 3C10C5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human