Order Entry
Denmark
ContactUsLinkComponent
 
Anti-ZBTB46 Rabbit Polyclonal Antibody
  NOVUNBP2-27211
undefined
Anti-ZBTB46 Rabbit Polyclonal Antibody
  NOVUNBP2-27211

 

  • Antibody type:
    Primary
  • Antigen name:
    zinc finger and BTB domain containing 46
  • Antigen symbol:
    ZBTB46
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Gene ID:
    140685
  • Antigen synonyms:
    BZEL|RINZF|BTBD4|dJ583P15.7|dJ583P15.8|ZNF340
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 μg
  • Pk:
    0,1 mg

 

 

Type: Primary
Antigen: ZBTB46 (zinc finger and BTB domain containing 46)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human