Anti-IKZF3 Rabbit Polyclonal Antibody
Katalog # NOVUNBP2-24495
Leverandør: Novus Biologicals
Specifikationer
- Antibody type:Primary
- Antigen name:IKAROS family zinc finger 3 (Aiolos)
- Antigen symbol:IKZF3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:22806
- Antigen synonyms:AIO|AIOLOS|ZNFN1A3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifikationer
Om denne vare
Type: Primary
Antigen: IKZF3 (IKAROS family zinc finger 3 (Aiolos))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: