Anti-GLI2 Rabbit Polyclonal Antibody (DyLight® 488)
Catalog # NOVUNBP2-23602G
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:GLI family zinc finger 2
- Antigen symbol:GLI2
- Clonality:Polyclonal
- Conjugation:DyLight® 488
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human,Mouse
- Environmentally Preferable:
- Gene ID:2736
- Antigen synonyms:THP2|HPE9|CJS|PHS2|THP1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: GLI2 (GLI family zinc finger 2)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse