Anti-ANXA1 Mouse Monoclonal Antibody [clone: 2F1]
Supplier: Novus Biologicals
Certificates
About this item
Type: Primary
Antigen: ANXA1 (annexin A1)
Clonality: monoclonal
Clone: 2F1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Specifications
- Catalog No:
- NOVUNBP2-23485SS
- NOVUNBP2-23485
- Antigen symbol:
- ANXA1
- ANXA1
- Antigen name:
- Annexin A1
- Annexin A1
- Antigen synonyms:
- ANX1|LPC1
- ANX1|LPC1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 2F1
- 2F1
- Host:
- Mouse
- Mouse
- Gene ID:
- 301
- 301
- Isotype:
- IgG1
- IgG1
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together