Order Entry
Denmark
ContactUsLinkComponent
 
Anti-LAMP2 Mouse Monoclonal Antibody [clone: H4B4]
Catalog # NOVUNBP2-22217
Supplier:  Novus Biologicals
undefined
Anti-LAMP2 Mouse Monoclonal Antibody [clone: H4B4]
Catalog # NOVUNBP2-22217
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    lysosomal-associated membrane protein 2
  • Antigen symbol:
    LAMP2
  • Clonality:
    Monoclonal
  • Clone:
    H4B4
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    3920
  • Antigen synonyms:
    LAMP-2|LGP110|LAMPB|CD107b
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 μL
  • Pk:
    0,1 mL

Specifications

About this item

Type: Primary
Antigen: LAMP2 (lysosomal-associated membrane protein 2)
Clonality: monoclonal
Clone: H4B4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human