Order Entry
Denmark
ContactUsLinkComponent
Anti-Glucose 6 Phosphate Dehydrogenase Mouse Monoclonal Antibody [clone: 2H7]
Anti-Glucose 6 Phosphate Dehydrogenase Mouse Monoclonal Antibody [clone: 2H7]
Supplier:  Novus Biologicals
Certificates
undefined
Anti-Glucose 6 Phosphate Dehydrogenase Mouse Monoclonal Antibody [clone: 2H7]
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNBP2-22125SS
  • NOVUNBP2-22125
  • Antigen symbol:
  • G6PD
  • G6PD
  • Antigen name:
  • Glucose-6-phosphate dehydrogenase
  • Glucose-6-phosphate dehydrogenase
  • Antigen synonyms:
  • G6PD1
  • G6PD1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • 2H7
  • 2H7
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 2539
  • 2539
  • Isotype:
  • IgG1
  • IgG1
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Frequently Bought Together