Anti-EBI3 Rabbit Polyclonal Antibody
NOVUNBP2-03944
- Antibody type:Primary
- Antigen name:Epstein-Barr virus induced 3
- Antigen symbol:EBI3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Mouse
- Western blot:Yes
- Gene ID:10148
- Antigen synonyms:IL35 subunit|IL-27B|EBV-induced gene 3 protein|IL27 subunit|Epstein-Barr virus induced gene 3|interleukin-27 subunit beta|cytokine receptor|IL-27 subunit beta|epstein-Barr virus-induced gene 3 protein|IL27B
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Rabbit Polyclonal EBI3 Antibody. Tested Applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Mouse, Rat.
Recommended Dilutions: Immunohistochemistry (Parafin): 5 ?g/ml
Type: Primary
Antigen: EBI3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse