Order Entry
Denmark
ContactUsLinkComponent
Anti-MMP7 Rabbit Polyclonal Antibody (DyLight® 405)
Anti-MMP7 Rabbit Polyclonal Antibody (DyLight® 405)
Catalog # NOVUNBP1-99123V
Supplier:  Novus Biologicals
Anti-MMP7 Rabbit Polyclonal Antibody (DyLight® 405)
Catalog # NOVUNBP1-99123V
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    matrix metallopeptidase 7 (matrilysin, uterine)
  • Antigen symbol:
    MMP7
  • Clonality:
    Polyclonal
  • Conjugation:
    DyLight® 405
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Human,
    Mouse
  • Environmentally Preferable:
  • Gene ID:
    4316
  • Antigen synonyms:
    PUMP-1|MPSL1|MMP-7
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 μL
  • Pk:
    0,1 mL

Specifications

About this item

Type: Primary
Antigen: MMP7 (matrix metallopeptidase 7 (matrilysin, uterine))
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse