Anti-DSG3 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 5H10]
Catalog # NOVUNBP1-78984H
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Desmoglein 3
- Antigen symbol:DSG3
- Clonality:Monoclonal
- Clone:5H10
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Gene ID:1830
- Antigen synonyms:CDHF6|PVA
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: DSG3 (desmoglein 3)
Clonality: monoclonal
Clone: 5H10
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human