Anti-DSG3 Mouse Monoclonal Antibody [clone: 5H10]
Supplier: Novus Biologicals
Certificates
About this item
The Desmoglein-3 Antibody (5H10) from Novus Biologicals is a mouse monoclonal antibody to Desmoglein-3. This antibody reacts with human. The Desmoglein-3 Antibody (5H10) has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:200
Type: Primary
Antigen: Desmoglein-3
Clonality: Monoclonal
Clone: 5H10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Specifications
- Catalog No:
- NOVUNBP1-78984
- NOVUNBP1-78984SS
- Antigen symbol:
- DSG3
- DSG3
- Antigen name:
- Desmoglein 3
- Desmoglein 3
- Antigen synonyms:
- 130 kDa pemphigus vulgaris antigen|130-kD pemphigus vulgaris antigen|CDHF6|Cadherin family member 6|desmoglein-3|PVA|pemphigus vulgaris antigen
- 130 kDa pemphigus vulgaris antigen|130-kD pemphigus vulgaris antigen|CDHF6|Cadherin family member 6|desmoglein-3|PVA|pemphigus vulgaris antigen
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 5H10
- 5H10
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 1830
- 1830
- Isotype:
- IgG1
- IgG1
- Western blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together