Anti-UBE2E1 Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
Certificates
About this item
The UbcH6 / UBE2E1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to UbcH6 / UBE2E1. This antibody reacts with human, mouse. The UbcH6 / UBE2E1 Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Western Blot (Negative).
Recommended Dilutions: Immunohistochemistry (Parafin): 1:600
Type: Primary
Antigen: UbcH6/UBE2E1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- NOVUNBP1-78983
- NOVUNBP1-78983SS
- Antigen symbol:
- UBCH6
- UBCH6
- Antigen name:
- Ubiquitin-conjugating enzyme E2E 1
- Ubiquitin-conjugating enzyme E2E 1
- Antigen synonyms:
- UBCH6
- UBCH6
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 7324
- 7324
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together