Anti-BNIP3L Rabbit Polyclonal Antibody (DyLight® 550)
NOVUNBP1-78264R
- Antibody type:Primary
- Antigen name:BCL2 Interacting Protein 3 like
- Antigen symbol:BNIP3L
- Clonality:Polyclonal
- Conjugation:DyLight® 550
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Mouse
- Gene ID:665
- Antigen synonyms:NIP3L|BNIP3A|BNIP3H|BNIP3a|Bnip3L|Adenovirus E1B19K-binding protein B5|NIX
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Type: Primary
Antigen: BNIP3L (BCL2/adenovirus E1B 19kDa interacting protein 3-like)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse