Anti-HSPA13 Rabbit Polyclonal Antibody (DyLight® 405)
NOVUNBP1-77455V
- Antibody type:Primary
- Antigen name:Heat shock 70kDA protein 1A
- Antigen symbol:HSPA1A
- Clonality:Polyclonal
- Conjugation:DyLight® 405
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human
- Gene ID:3303
- Antigen synonyms:HSPA1|HSP70-1A|HSP72|HSP70-1|HSP70.1|HEL-S-103|HSP70I
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Type: Primary
Antigen: HSPA13 (heat shock protein 70kDa family, member 13)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human