Anti-HSPD1 Rabbit Polyclonal Antibody (DyLight® 650)
Catalog # NOVUNBP1-77396C
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Heat shock 60kDa protein 1
- Antigen symbol:HSPD1
- Clonality:Polyclonal
- Conjugation:DyLight® 650
- Host:Rabbit
- ImmunoFluorescence:Yes
- Western blot:Yes
- Gene ID:3329
- Antigen synonyms:GROEL|HSP65|HLD4|HSP-60|HuCHA60|Chaperonin 60|HSP60|Mitochondrial Matrix Protein P1|CPN60|Heat Shock Protein 65|SPG13
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: HSPD1 (heat shock 60kDa protein 1 (chaperonin))
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: