Order Entry
Denmark
ContactUsLinkComponent
Anti-CHD7 Rabbit Polyclonal Antibody (Biotin)
Anti-CHD7 Rabbit Polyclonal Antibody (Biotin)
Catalog # NOVUNBP1-77393B
Supplier:  Novus Biologicals
undefined
Anti-CHD7 Rabbit Polyclonal Antibody (Biotin)
Catalog # NOVUNBP1-77393B
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Chromodomain helicase DNA binding protein 7
  • Antigen symbol:
    CHD7
  • Clonality:
    Polyclonal
  • Conjugation:
    Biotin
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Gene ID:
    55636
  • Antigen synonyms:
    HH5|KAL5|ATP-dependent helicase CHD7|chromodomain helicase DNA binding protein 7 isoform CRA_e|IS3|chromodomain-helicase-DNA-binding protein 7
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µL
  • Pk:
    0,1 mL

Specifications

About this item

Type: Primary
Antigen: CHD7 (chromodomain helicase DNA binding protein 7)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse