Anti-GABARAP Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
Certificates
About this item
The GABARAP Antibody from Novus Biologicals is a rabbit polyclonal antibody to GABARAP. This antibody reacts with human, mouse. The GABARAP Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:200
Type: Primary
Antigen: GABARAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- NOVUNBP1-71771
- NOVUNBP1-71771SS
- Antigen symbol:
- GABARAP
- GABARAP
- Antigen name:
- Gamma-aminobutyric acid receptor-associated protein
- Gamma-aminobutyric acid receptor-associated protein
- Antigen synonyms:
- GABARAP-a|MM46|GABA(A) receptor-associated protein|FLC3B|ATG8A|Gamma-aminobutyric acid receptor-associated protein
- GABARAP-a|MM46|GABA(A) receptor-associated protein|FLC3B|ATG8A|Gamma-aminobutyric acid receptor-associated protein
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 11337
- 11337
- Western blot:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together