Anti-PTCH1 Rabbit Polyclonal Antibody (DyLight® 550)
Catalog # NOVUNBP1-71662R
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Patched 1
- Antigen symbol:PTCH1
- Clonality:Polyclonal
- Conjugation:DyLight® 550
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:5727
- Antigen synonyms:etID309849.2|Patched1|PTC-1|Patched 1 |PTC1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: PTCH1 (patched 1)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human