Order Entry
Denmark
ContactUsLinkComponent
Anti-GPR155 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anti-GPR155 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # NOVUNBP1-50626H
Supplier:  Novus Biologicals
Anti-GPR155 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # NOVUNBP1-50626H
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    G Protein-Coupled Receptor 155
  • Antigen symbol:
    GPR155
  • Clonality:
    Polyclonal
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    151556
  • Antigen synonyms:
    PGR22|DEPDC3|DEP.7
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µL
  • Pk:
    0,1 mL

Specifications

About this item

Type: Primary
Antigen: GPR155 (G protein-coupled receptor 155)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human