Order Entry
Denmark
ContactUsLinkComponent
Anti-N Cadherin Mouse Monoclonal Antibody (DyLight® 488) [clone: 13A9]
Anti-N Cadherin Mouse Monoclonal Antibody (DyLight® 488) [clone: 13A9]
Catalog # NOVUNBP1-48309G
Supplier:  Novus Biologicals
Anti-N Cadherin Mouse Monoclonal Antibody (DyLight® 488) [clone: 13A9]
Catalog # NOVUNBP1-48309G
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cadherin 2
  • Antigen symbol:
    CDH2
  • Clonality:
    Monoclonal
  • Clone:
    13A9
  • Conjugation:
    DyLight® 488
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    1000
  • Antigen synonyms:
    CDHN|Neural-Cadherin|CDw325|N-Cadherin|CDH2|NCAD
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µL
  • Pk:
    0,1 mL

Specifications

About this item

Type: Primary
Antigen: N Cadherin
Clonality: monoclonal
Clone: 13A9
Conjugation: DyLight® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat