Order Entry
Denmark
ContactUsLinkComponent
 
Anti-N Cadherin Mouse Monoclonal Antibody (DyLight® 680) [clone: 13A9]
Catalog # NOVUNBP1-48309FR
Supplier:  Novus Biologicals
Anti-N Cadherin Mouse Monoclonal Antibody (DyLight® 680) [clone: 13A9]
Catalog # NOVUNBP1-48309FR
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    cadherin 2, type 1, N-cadherin (neuronal)
  • Antigen symbol:
    CDH2
  • Clonality:
    Monoclonal
  • Clone:
    13A9
  • Conjugation:
    DyLight® 680
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Gene ID:
    1000
  • Antigen synonyms:
    CDHN|CD325|CDw325|NCAD
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 Tests
  • Pk:
    100 Tests

Specifications

About this item

Type: Primary
Antigen: N Cadherin
Clonality: monoclonal
Clone: 13A9
Conjugation: DyLight® 680
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat