Anti-LOX propeptide Rabbit Polyclonal Antibody (DyLight® 405)
NOVUNBP1-30327V
- Antibody type:Primary
- Antigen name:lysyl oxidase
- Antigen symbol:LOX
- Clonality:Polyclonal
- Conjugation:DyLight® 405
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human,Rat
- Western blot:Yes
- Gene ID:4015
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Type: Primary
Antigen: LOX propeptide
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Rat