Order Entry
Denmark
ContactUsLinkComponent
Anti-BMP2 Rabbit Polyclonal Antibody
Anti-BMP2 Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals
Certificates
Anti-BMP2 Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNBP1-19751SS
  • NOVUNBP1-19751
  • Antigen symbol:
  • BMP2
  • BMP2
  • Antigen name:
  • Pregnancy specific beta-1-glycoprotein 4
  • Pregnancy specific beta-1-glycoprotein 4
  • Antigen synonyms:
  • BMP2A|BDA2
  • BMP2A|BDA2
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 650
  • 650
  • Western blot:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together