Anti-BHLHE41 Rabbit Polyclonal Antibody
NOVUNBP1-19613
- Antibody type:Primary
- Antigen name:basic helix-loop-helix family, member e41
- Antigen symbol:SHARP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:79365
- Antigen synonyms:BHLHB3|DEC2|hDEC2|SHARP1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
The DEC2 / SHARP1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to DEC2 / SHARP1. This antibody reacts with human, mouse, rat. The DEC2 / SHARP1 Antibody has been validated for the following applications: Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:50-1:200
Type: Primary
Antigen: DEC2/SHARP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat