Anti-PFN1 Rabbit Polyclonal Antibody (DyLight® 488)
NOVUNBP1-19344G
- Antibody type:Primary
- Antigen name:Profilin 1
- Antigen symbol:PFN1
- Clonality:Polyclonal
- Conjugation:DyLight® 488
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:5216
- Antigen synonyms:ALS18
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Type: Primary
Antigen: PFN1 (profilin 1)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Bovine, Human, Mouse, Rat