Anti-GLO1 Mouse Monoclonal Antibody (DyLight® 488) [clone: Glo1a]
Catalog # NOVUNBP1-19015G
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Glyoxalase 1
- Antigen symbol:GLO1
- Clonality:Monoclonal
- Clone:Glo1a
- Conjugation:DyLight® 488
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Gene ID:2739
- Antigen synonyms:Glo1-s|Glo1-r|AW550643|GLYI|0610009E22Rik|Glo-1r|GLY1|Glo-1s|1110008E19Rik|Qglo|HEL-S-74|2510049H23Rik|Glo-1|GLOD1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: GLO1 (glyoxalase I)
Clonality: monoclonal
Clone: Glo1a
Conjugation: DyLight® 488
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse