Anti-HPi3 Mouse Monoclonal Antibody (DyLight® 550) [clone: HIC1-7H10]
Catalog # NOVUNBP1-18947R
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Heterochromatin Protein 3
- Antigen symbol:HPI3
- Clonality:Monoclonal
- Clone:HIC1-7H10
- Conjugation:DyLight® 550
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: HPi3
Clonality: monoclonal
Clone: HIC1-7H10
Conjugation: DyLight® 550
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human