Order Entry
Denmark
ContactUsLinkComponent
Anti-HPi3 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: HIC1-7H10]
Anti-HPi3 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: HIC1-7H10]
Catalog # NOVUNBP1-18947APC
Supplier:  Novus Biologicals
Anti-HPi3 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: HIC1-7H10]
Catalog # NOVUNBP1-18947APC
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    HPI3
  • Clonality:
    Monoclonal
  • Clone:
    HIC1-7H10
  • Conjugation:
    APC (Allophycocyanin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 Tests
  • Pk:
    100 Tests

Specifications

About this item

Type: Primary
Antigen: HPi3
Clonality: monoclonal
Clone: HIC1-7H10
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human