Order Entry
Denmark
ContactUsLinkComponent
Anti-AGR2 Rabbit Polyclonal Antibody (Biotin)
Anti-AGR2 Rabbit Polyclonal Antibody (Biotin)
  NOVUNBP1-05936B
undefined
Anti-AGR2 Rabbit Polyclonal Antibody (Biotin)
  NOVUNBP1-05936B

 

  • Antibody type:
    Primary
  • Antigen name:
    Anterior Gradient 2
  • Antigen symbol:
    AGR2
  • Clonality:
    Polyclonal
  • Conjugation:
    Biotin
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Gene ID:
    10551
  • Antigen synonyms:
    AG2|hAG-2|AG-2|Anterior gradient protein 2 homolog|HPC8|Secreted cement gland protein XAG-2 homolog
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µL
  • Pk:
    0,1 mL

 

 

The AG-2 Antibody [Biotin] from Novus Biologicals is a rabbit polyclonal antibody to AG-2. This antibody reacts with human, mouse, bovine. The AG-2 Antibody [Biotin] has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:100

Type: Primary
Antigen: AG-2
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Bovine