Anti-SLC2A9 Rabbit Polyclonal Antibody (DyLight® 488)
Catalog # NOVUNBP1-05054G
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Glucose Transporter 9
- Antigen symbol:GLUT9
- Clonality:Polyclonal
- Conjugation:DyLight® 488
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:56606
- Antigen synonyms:GLUTX|UAQTL2|GLUT9|URATv1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: SLC2A9 (solute carrier family 2 (facilitated glucose transporter), member 9)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse