Anti-SIGLEC1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: HSn 7D2]
Catalog # NOVUNB600-534F
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Sialic acid binding Ig-like lectin 1, sialoadhesin
- Antigen symbol:SIGLEC1
- Clonality:Monoclonal
- Clone:HSn 7D2
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Gene ID:6614
- Antigen synonyms:dJ1009E24.1|SN|CD169|SIGLEC-1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: SIGLEC1 (sialic acid binding Ig-like lectin 1, sialoadhesin)
Clonality: monoclonal
Clone: HSn 7D2
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human