Order Entry
Denmark
ContactUsLinkComponent
Anti-WNK4 Rabbit Polyclonal Antibody (DyLight® 488)
Anti-WNK4 Rabbit Polyclonal Antibody (DyLight® 488)
Catalog # NOVUNB600-284G
Supplier:  Novus Biologicals
Anti-WNK4 Rabbit Polyclonal Antibody (DyLight® 488)
Catalog # NOVUNB600-284G
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    WNK lysine deficient Protein kinase 4
  • Antigen symbol:
    WNK4
  • Clonality:
    Polyclonal
  • Conjugation:
    DyLight® 488
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    65266
  • Antigen synonyms:
    PHA2B|PRKWNK4
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µL
  • Pk:
    0,1 mL

Specifications

About this item

Type: Primary
Antigen: WNK4 (WNK lysine deficient protein kinase 4)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat