Anti-BIRC5 Mouse Monoclonal Antibody (DyLight® 350) [clone: 32.1]
Catalog # NOVUNB500-237UV
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:baculoviral IAP repeat containing 5
- Antigen symbol:BIRC5
- Clonality:Monoclonal
- Clone:32.1
- Conjugation:DyLight® 350
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Western blot:Yes
- Gene ID:332
- Antigen synonyms:API4|EPR-1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 Tests
- Pk:100 Tests
Specifications
About this item
Type: Primary
Antigen: BIRC5 (baculoviral IAP repeat containing 5)
Clonality: monoclonal
Clone: 32.1
Conjugation: DyLight® 350
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: