Anti-BIRC5 Mouse Monoclonal Antibody (DyLight® 488) [clone: 32.1]
Catalog # NOVUNB500-237G
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Baculoviral IAP Repeat Containing 5
- Antigen symbol:BIRC5
- Clonality:Monoclonal
- Clone:32.1
- Conjugation:DyLight® 488
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Gene ID:332
- Antigen synonyms:Apoptosis Inhibitor 4|SVV|TIAP|IAP4|API4|Baculoviral IAP repeat containing 5|EPR 1|Apoptosis inhibitor survivin|BIRC 5
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: BIRC5 (baculoviral IAP repeat containing 5)
Clonality: monoclonal
Clone: 32.1
Conjugation: DyLight® 488
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: