Anti-NUMB Rabbit Polyclonal Antibody (DyLight® 405)
Catalog # NOVUNB500-178V
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:numb homolog (Drosophila)
- Antigen symbol:NUMB
- Clonality:Polyclonal
- Conjugation:DyLight® 405
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Chicken,Human,Mouse
- Western blot:Yes
- Gene ID:8650
- Antigen synonyms:S171|C14orf41|c14_5527
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: NUMB (numb homolog (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Chicken, Human, Mouse